Tris HCl Buffer Solutions 1.0 M, Rockland
Supplier: Rockland Immunochemical
Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
These solutions were aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The products were tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: These products ares for research use only and are not intended for therapeutic or diagnostic applications.