Fritted Empty Cartridges for Solid Sample Loading, Bonna Agela Technologies
Supplier: Phenomenex
FCH004-H FCH012-H FCH020-H FCH040-H FCH080-H FCH120-H FCH330-H FCH800-H FCH1500-H FCH080-S FCH120-S
89214-206PK 296.08 CAD
89214-206 89214-208 89214-210 89214-212 89214-214 89214-216 89214-218 89214-220 89214-222 89238-004 89238-006
Fritted Empty Cartridges for Solid Sample Loading, Bonna Agela Technologies
Chromatography Columns
These empty flash columns contain one column, one cap and two frit plates.