Uvex Tirade™ Sealed Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
S4044 S4041 S4042 S4043
10790-600CS 6906.33 CAD
10790-600 10790-682 10790-684 10790-686
Uvex Tirade™ Sealed Eyewear, Honeywell Safety
Safety Glasses
Uvex Tirade™ sealed eyewear offers protection against impact, wind, airborne debris, sunlight and glare.
  • Hypoallergenic latex-free option available
  • 100% non-conductive dielectric design
  • Dependable, cost-effective solution for indoor and outdoor environments requiring sealed eyewear protection
  • Uvextra® anti-fog coated lens reduces fogging, keeping lenses clear
  • Removable temples and standard adjustable headband also available
  • Can be worn in environments where accidental exposure to electrical charge is possible
Uvex Tirade™ features a large unilens design for a wide field of unobstructed peripheral vision, and a soft foam subframe to ensure a comfortable gap-free fit.
Certifications: Meets ANSI Z87.1-2010 and is certified to the requirements of the CSA Z94.3 standards (with and without foam subframe in place).
Delivery information: All Uvex Tirade™ are supplied with temples and headband as well as with instructions and warnings for proper use.