Chem Snap Caps™, Colour-Coded, Chemglass
Supplier: Chemglass
CV-3500-R011 CV-3501-0011 CV-3550-0011 CV-3550-G011 CV-3570-0011 CV-3570-B011 CV-3590-0011 CV-3590-B011 CV-3590-G011 CV-3630-0011 CV-3630-S011
75820-768PK 25.7 CAD
75820-768 75820-772 75820-774 75820-778 75820-784 75820-786 75820-804 75820-806 75820-808 75820-830 75820-840
Chem Snap Caps™, Colour-Coded, Chemglass
Closures Test/Sample Tube Closures
These 11mm Snap Caps™ can be snapped on and off by hand, eliminating the need for metal crimping and decapping tools, and reducing potential hazards due to sharp aluminum edges.
  • Large opening provides greater target area for needle penetration
  • Color coded for easy sample identification
These liner-less Chem Snap Caps™ are available with or without starburst. The starburst allows for easier needle penetration. These caps were developed to handle the most volatile sample providing a liquid airtight seal to the vial. By snapping on the cap, anMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable