Magid® Gemstone Diamond Protective Visitor Glasses, Magid Glove
Supplier: Magid Glove
Y20CHC Y20C
62300-100CS 433.13 CAD
62300-100 62300-112
Magid® Gemstone Diamond Protective Visitor Glasses, Magid Glove
Safety Glasses
These glasses feature a contemporary wrap-around frame with a clear polycarbonate lens.
  • Visitor spec
  • Vented sideshields
  • UV protective glasses fit comfortably over prescription glasses
The Y20C spectacle also utilizes an impact-resistant lens. Glasses provide 99.9% UV protection.
Certifications: Meet ANSI Z87.1-2010 High Impact Standards.
Packaging: Each pair is wrapped in a sealed polybag.