Puritan® Lim Broth Transport Medium, Puritan Medical Products
Supplier: Puritan Medical Products
LB-200
LB-500
75998-434CS
173.49
CAD
75998-434
75998-436
Puritan® Lim Broth Transport Medium, Puritan Medical Products
Media for Microbiology
Prepared Media for Microbiology
Selective enrichment medium for use in the qualitative procedures for the isolation of Group B Streptococcus (GBS) from clinical specimens.
- 2 mL or 5 mL Lim Broth Selective Enrichment Medium
- For In Vitro Diagnostic Use
- Light blue cap
For optimum performance, store at 2 to 25°C.
Certifications: Made in USA.
Ordering information: Products are final sale.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...