KIMAX® KimCote® Culture Tubes, Reusable, with Unattached Caps, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
KC45066-13100 KC45066-16125 KC45066-20150 KC45066-25150
89097-440CS 1291.29 CAD
89097-440 89097-442 89097-444 89097-446
KIMAX® KimCote® Culture Tubes, Reusable, with Unattached Caps, Kimble Chase
Tubes Culture Tubes
KimCote® plastic-coated culture tubes are made from tubing with uniform wall thickness for maximum heat transfer and chemical resistance.
  • Ideal for biohazard test protocols
  • Supplied with phenolic screw-thread caps, unattached
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The safety coating reduces the hazards of shattered glass and leakage of toxic or hazardous materials.
Certifications: ASTM E982, Type VI, Class A requirements.