Clonetics™ OGM™ Osteoblast Growth Medium, BulletKit, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-4193 CC-4194 CC-3207
CA10063-996EA 481.9 CAD
CA10063-996 CA10063-998 CA10065-294
Clonetics™ OGM™ Osteoblast Growth Medium, BulletKit, Lonza
Cell Culture Media
Clonetics™ OGM™ Osteoblast Growth Medium is specially formulated for the growth and expansion of human osteoblasts.
It contains 10% serum. OGM™ Differentiation Supplements are recommended for differentiation. It supports the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.