POWERCOAT® Disposable Tri-polymer Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
PSD-TRIP-S PSD-TRIP-M PSD-TRIP-XL
CA56619-976CS 611.88 CAD
CA56619-976 CA56619-978 CA56619-980 CA56619-982
POWERCOAT® Disposable Tri-polymer Gloves, Honeywell Safety
Gloves
These are tri-polymer disposable gloves with a blend of latex, neoprene and nitrile that provides excellent chemical and liquid protection.
  • Powder-free for reduced product contamination
  • Finger tips have a rough finish for added grip in wet applications
Ideal for use in agriculture, chemical, oil and gas industries.
Certifications: According to ANSI.