Ashby’s Mannitol Agar, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M706-500G
M706-2.5KG
M706-5KG
CA61000-154EA
1320.26
CAD
CA61000-154
CA89226-212
CA89226-214
Ashby’s Mannitol Agar, HiMedia Laboratories
Media for Microbiology
Dehydrated Media for Microbiology
Ashby's mannitol agar is used for cultivation of Azotobacter species that can use mannitol and atmospheric nitrogen as source of carbon and nitrogen respectively.
- White to cream homogeneous free flowing powder
- Firm, comparable with 1.5% Agar gel