HiVeg™ Staphylococcus Agar, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV156-500G M156-500G M156-2.5KG M156-5KG
CA61000-692EA 1505.99 CAD
CA61000-692 CA95021-280 CA89224-276 CA89224-278
HiVeg™ Staphylococcus Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Staphylococcus Agar No.110 is used as a selective medium for the isolation and testing of pathogenic Staphylococci.