Airgas® Manifold Protocol Station
Supplier: Airgas
Y15-12CMH320 Y15-12CMH326 Y15-12CMH346 Y15-12CMH350 Y15-12CMH540 Y15-12CMH580 Y15-12CMH590
47752-544EA 4433.83 CAD
47752-544 47752-546 47752-548 47752-550 47752-552 47752-554 47752-556
Airgas® Manifold Protocol Station
Manifolds
These two-station manifolds are designed to provide a safe, reliable method of wall-mounting a single cylinder regulator, eliminating the need to handle the regulator during cylinder change out.
  • Made from 304 stainless steel
  • Durable, long-life service
  • Easily mount to wall or existing structure
Manifolds are provided with 36" stainless steel (corrugat-ed bellows) flexible pigtails and CGA connections with check valves to prevent contaminates from entering during cylinder change-out.