Guanosine 5'-triphosphate disodium salt ≥90% (by HPLC), white to off-white powder

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: ((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Guanosine-5'-triphosphate disodium salt, GTP disodium salt, Guanosine-5-triphosphoric aicd disodium salt

00348-1G 00348-5G
CH00348-1GEA 86.9 CAD
CH00348-1G CH00348-5G
Guanosine 5'-triphosphate disodium salt ≥90% (by HPLC), white to off-white powder
Guanosine 5'-triphosphate disodium salt

  • Synonyms: 5-GTP.2Na

Formula: C₁₀H₁₄N₅Na₂O₁₄P₃
MW: 567.147 g/mol
MDL Number: MFCD03410297
CAS Number: 56001-37-7

Order Now

Specification Test Results

Appearance White to off-white powder
Soluble in water Passes test
Ethanol or ether Partially insoluble
pH 3.0 - 5.0
UV Absorbance A250/A260 0.92 to 1.00
UV Absorbance A280/A260 0.65 to 0.69
Loss on Drying ≤15%
Assay (anhydrous basis) ≥90%
Purity by HPLC ≥90%
Storage ≤-4°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR