L(-)-Tryptophan ≥99%, crystalline powder

Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH

00300-100G 00300-250G 00300-25G
CH00300-100GEA 74.21 CAD
CH00300-100G CH00300-250G CH00300-25G
L(-)-Tryptophan ≥99%, crystalline powder
L(-)-Tryptophan

  • Synonyms: L-Trp-OH; (S)-2-Amino-3-(3-indolyl)propionic acid

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 290 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
Merck Index: 12,09929

Order Now

Specification Test Results

Assay 98.5-101.5%
Appearance White to light yellow crystalline powder
Heavy Metals (as Pb) ≤ 20 ppm
Arsenic (As₂O₂) ≤1 ppm
Chloride (Cl-) ≤0.02%
Sulfate (SO₄²⁻) ≤ 0.03%
Loss on Drying ≤0.30%
Residue On Ignition ≤0.10%
Optical Rotation -30.0 to -33.0°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR