Pierce-It™ Closures, Simport
Supplier: Simport Scientific
T407-16L T407-16R T407-16Y
10127-374CS 184.54 CAD
10127-374 10127-376 10127-380
Pierce-It™ Closures, Simport
Closures Test/Sample Tube Closures
Made of thermoplastic elastomer, these disposable closures produce a firm, leak-resistant seal for glass and plastic tubes
  • The flexible plastic cap can be pierced as often as needed
  • Will not interfere with most common chemistry, coagulation, and drug monitoring methodologies
  • Can be stored in the refrigerator, in the freezer, or at room temperature
  • Withstand agitation in a test tube Vortex mixer
  • Available in 8 colors for easy identification
Two sizes, 13mm and 16mm, fit a variety of tubes including glass evacuated blood drawing tubes. Will also fit 12More Product Information