Cleanert® Alumina N SPE Columns, Bonna-Agela Technologies
Supplier: Phenomenex
AL0006-N AL1001-N AL2003-N AL5003-N AL5006-N
97017-902PK 426.28 CAD
97017-902 97017-906 97017-910 97017-912 97017-914
Cleanert® Alumina N SPE Columns, Bonna-Agela Technologies
Chromatography Columns
Cleanert® Alumina N sorbent can adsorb molecules by interaction with the aluminum metal center.
The neutralized surface allows interaction with compounds whose heteroatoms are electronegative or with an electron-rich highly aromatic structure.This material is generally used in the detection of pigment such as Sudan and malachite green.
Order Now
SPECIFICATIONS
Environmentally Preferable