Sodium chloride 0.9% in aqueous solution endotoxin-free, sterile

Supplier: ADIPOGEN CORP MS

IAX-900-003-LD15 IAX-900-003-3010
102979-272EA 49.23 CAD
102979-272 102979-276
Sodium chloride 0.9% in aqueous solution endotoxin-free, sterile
Sodium chloride

Use for the dilution of TLRpure LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).

Formula: NaCl
Storage Temperature: Refrigerator
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Formulation Liquid. 1.5ml (IAX-900-003-LD15) or 10ml (IAX-900-003-L010) Physiological Saline [Sodium Chloride 0.9%] in sterile filtered double-distilled water (ddWater), without any additives.
Endotoxin <0.0002EU/µg, after repeated purification cycles over activated charcoal.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR