PBS Endotoxin-free (sterile), AdipoGen Life Sciences

Supplier: ADIPOGEN CORP MS

IAX-900-001-LD15 IAX-900-001-L010 IAX-900-001-3010
102979-260EA 49.23 CAD
102979-260 102979-262 102979-264
PBS Endotoxin-free (sterile), AdipoGen Life Sciences
Buffers Cell Culture Buffers

Endotoxin free buffer used for the dilution of TLRpure LPS and solubilisation of endotoxin-free reagents (e.g


Formulation: Liquid. 1.5ml (IAX-900-001-LD15) or 10ml (IAX-900-001-L010) PBS pH 7.2 in sterile filtered double-distilled water (ddWater), without any additives.


Endotoxin: <0.0002EU/µg, after repeated purification cycles over activated charcoal.


ODNs).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR