ZnAF-1F DA ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS

CDX-Z0011-M001 CDX-Z0011-M005
102981-260EA 266.15 CAD
102981-260 102981-262
ZnAF-1F DA ≥95% (by HPCE)
ZnAF-1F DA

For application to living cells, the diacetyl derivative of ZnAF-1F, ZnAF-1F DA, was synthesized. ZnAF-1F DA can permeate through the cell membrane, and is hydrolyzed by esterase in the cytosol to yield ZnAF-1F, which is retained in the cells. ZnAF-1F DA has been used to measure the changes of intracellular Zn2+ in cultured cells and hippocampal slices. Spectral data: lambdaex 492nm; lambdaem 514nm in PBS.

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR