ZnAF-1 DA ≥90% (by HPCE)

Supplier: ADIPOGEN CORP MS

CDX-Z0007-M001 CDX-Z0007-M005 CDX-Z0007-M025
102981-254EA 218.46 CAD
102981-254 102981-256 102981-258
ZnAF-1 DA ≥90% (by HPCE)
ZnAF-1 DA

Suitable for the fluorimetric detection of Zn2+ (after esterase hydrolysis). Spectral data: lambdaex 492nm; lambdaem 515nm in PBS.

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >90% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR