Starch, red coloured
Supplier: ADIPOGEN CORP MS
CDX-S0035-G001
CDX-S0035-G005
102981-246EA
241.54
CAD
102981-246
102981-248
Starch, red coloured
Starch
Red-dyed soluble starch. An excellent soluble substrate for alpha-amylase assay activity. Used for the measurement of enzyme activity, for research, forensic research, biochemical enzyme assays and in vitro diagnostic analysis.
- Synthetic
- Soluble in 0.5M potassium chloride solution
Formula:
(C₆H₁₀O₅)n Storage Temperature: Freezer |
MDL Number:
MFCD00082026 CAS Number: 9005-25-8 |
Specification Test Results
Source/Host | Synthetic |
Appearance/Color | Red or pink powder. |
Solubility | Soluble in 0.5M potassium chloride solution. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...