Red arabinan
Supplier: ADIPOGEN CORP MS
CDX-R0054-M500
CDX-R0054-G001
102981-242EA
183.08
CAD
102981-242
102981-244
Red arabinan
Red arabinan
Red-dyed arabinan. Soluble substrate for endo-1,5-alpha-L-arabinanase assay activity. Used for the measurement of enzyme activity, for research, biochemical enzyme assays and in vitro diagnostic analysis.
Specification Test Results
Source/Host | Synthetic |
Appearance/Color | Pink to purple red powder. |
Solubility | Soluble in water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...