DAR-4T ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-D0275-M001
CDX-D0275-M005
102981-232EA
484.62
CAD
102981-232
102981-234
DAR-4T ≥98% (by NMR)
DAR-4T
Reference compound for the fluorescent (nitric oxide) NO probe DAR-4.
Specification Test Results
Source/Host | Synthetic |
Purity | >98% (NMR) |
Appearance/Color | Dark violet powder. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...