Azocarob galactomannan
Supplier: ADIPOGEN CORP MS
|
Danger
|
CDX-A0221-M500
CDX-A0221-G001
102981-228EA
183.08
CAD
102981-228
102981-230
Azocarob galactomannan
Azocarob galactomannan
Remazol brilliant blue-dyed galactomannan polysaccharide. Soluble substrate for the specific measurement of endo-1,4-beta-D-mannanase/galactomannase. Used for the measurement of enzyme activity, for research, biochemical enzyme assays and in vitro diagnostic analysis.
Specification Test Results
Source/Host | Synthetic |
Appearance/Color | Dark blue powder. |
Solubility | Soluble in water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...