Aftin-5 ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (2R)-2-[[9-Isopropyl-6-(N-methylanilino)purin-2-yl]amino]butan-1-ol, Amyloidβ Forty-Two Inducer 5

AG-MR-C0015-M001 AG-MR-C0015-M005 AG-MR-C0015-M025
102981-184EA 130.77 CAD
102981-184 102981-186 102981-188
Aftin-5 ≥98%
Aftin-5

Roscovitine-related purine with no activity on CDKs (used as control for roscovitine). Selectively and potently increases production of extracellular Abeta42 and decreases production of extracellular Abeta38 in cultured cells. Extracellular Abeta40 levels remain stable. Intracellular levels of these amyloids appear to remain stable. Alzheimer's Disease (AD) accelerator that interacts with VDAC1, prohibitin and mitofilin, possibly interfering with subcellular compartmentalization and lipid rafts properties, shifting gamma-secretase activity toward Abeta42 generation. Induces a reversible mitochondrial phenotype reminiscent of the one observed in AD brains. Tool to detect inhibitors of Aftin-induced actions (potential anti-AD compounds).

Formula: C19H26N6O
MW: 354.5 g/mol
Storage Temperature: Freezer
CAS Number: 1461717-05-4

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR