IHR-NAc ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-3527-M001 AG-CR1-3527-M005
102981-176EA 115.38 CAD
102981-176 102981-178
IHR-NAc ≥98% (by NMR)
IHR-NAc

Sonic hedgehog (Shh) signaling pathway inhibitor. Potential anticancer compound. Cell membrane permeable Smoothened (Smo) antagonist. Acts on extra- and intracellular exposed Smo. Binds to the seven-transmembrane (7TM) pocket. Blocks hedgehog-induced movement of Smo into the primary cilium, disengages Smo-dependent abrogation of Gli2 and Gli3 proteolytic processing and inhibits Gli activity induced by loss of PTCH1.

Order Now

Specification Test Results

Purity >98% (NMR)
Appearance/Color White to grey solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR