Nalgene® Certified Clean Polycarbonate Biotainer™ Carboys, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
383405-16 383405-42 383410-08 383410-42 383423-42
75832-264CS 950.53 CAD
75832-264 75832-266 75832-268 75832-270 75832-272
Nalgene® Certified Clean Polycarbonate Biotainer™ Carboys, Sterile, Thermo Scientific
Bottles Carboys
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific Nalgene Polycarbonate Biotainer Bottles and Carboys reduce the risk of carry-over contamination.
  • Sterile and ready to use
  • Translucent construction
  • Robust and versatile
Durable, translucent PC construction ensures the integrity of your sample, even frozen to −100 °C. Sterile, ready-to-use products with a Sterility Assurance Level of 10⁻⁶ inside the bottle/closure. Robust, PC can be used from −100 to 100 °C, allowing for both heated mixing and frozen storage or shipment. Must be tested in actual use conditions as results may vary depending upon application. Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications. All container/closure systems are certified leakproof to protect valuable contents.
Certifications: Certified clean products, which meet USP <788> standards and endotoxins below 0.5 EU/ml.
Ordering information: Products can be supplied with forced extraction studies, validation binders and certificates.
Additionally our certified clean products include clean room produced in ISO Class 7 to limit particulates during molding; two heat-sealed sterile barriers to protect product prior to use; heat-sealed bags include quick-tears on the top and bottom and placed in carton liner twisted and tape/tie sealed to prevent gross contamination on the inner packaging; certified and lot-to-lot tested to meet USP <788> low particulate requirements to limit potential particulate contamination; certified low endotoxin (0.50 EU/ml) according to USP <85> to reduce pyrogens.