Hydrochloric acid solution 6.0 N, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = R
BAKR0347-02
BAKR0347-07
CAJT0347-2CS
15311.81
CAD
CAJT0347-2
CAJT0347-07
Hydrochloric acid solution 6.0 N, Biotech Reagent, J.T.Baker®
Hydrochloric acid
Made from USP Water for injection (WFI) and Hydrochloric acid, NF.
Formula:
HCl Density: 1.06 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,II |
Specification Test Results
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Appearance (Clear, colorless liquid) | Passes Test |
Normality | 5.95 - 6.05 |
Ammonium (NH₄) | ≤ 3 ppm |
Trace Impurities - Heavy Metals | ≤ 0.5 ppm |
Trace Impurities - Iron (Fe) | ≤ 0.1 ppm |
Sulfate (SO₄) | ≤ 1 ppm |
Made from U.S.P. Water for Injection (WFI) and Hydrochloric Acid, N.F. | |
(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical | |
Specifications | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Filtered through a 0.2 micron filter. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...