Radioactive Source, Set of 8
Supplier: SPECTRUM TECHNIQUES, LLC
RSS8EU RSS8UN RSS8
470220-464PK 872.25 CAD
470220-464 470220-512 470220-514 470337-082 470337-083
Radioactive Source, Set of 8
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Convenient set of 8 for studying gamma spectroscopy.
  • Safe to use
  • Easy disposal
  • All Gamma emissions
This set contains eight different gamma emitting isotopes covering the entire energy range from 32 to 1333 keV. Also included in the set is a mixed source of Cs-137 and Zn-65 which students may use to identify an “unknown” isotope.
The set consists of Ba-133, Cd-109, Co-57, Co-60, Cs-137, Mn-54. Na-22 and Cs/Zn.
Source activities are all 1 uCi, except the Cs which is 0.5 μCi and Zn which is 1 μCi Zn.
470220-464 is the same set except the activity ofMore Product Information