GW806742X ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1215-M005 SYN-1215-M010
102979-862EA 393.85 CAD
102979-862 102979-864
GW806742X ≥95%
GW806742X

Potent protein kinase VEGFR2 inhibitor (2nm range). Binds to the pseudokinase domain of MLKL and blocks cell death by necroptosis, as well. With regards to its role in necroptosis, it is an ATP-mimetic that was shown to bind recombinant mouse MLKL pseudokinase domain, which showed inhibited TSQ-induced death of mouse dermal fibroblasts by delaying MLKL translocation to the membrane. Appears therefore to be a valuable reagent to inhibit necroptosis. It also provides an important proof-of-principle that targeting catalytically-dead pseudoenzymes represents a feasible, emerging therapeutic avenue.

Formula: C₂₅H₂₂F₃N₇O₄S
Storage Temperature: Freezer
MDL Number: MFCD29036385
CAS Number: 579515-63-2

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR