Kilo Scale Thermocouple Probe Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5279-03 5279-10
75877-538EA 867.92 CAD
75877-538 75877-540
Kilo Scale Thermocouple Probe Adapter, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
Anti-whip, glass thermocouple probe adapter for protecting ¼" O.D
  • Heavy borosilicate glass
  • Available as complete unit or glass sheath only
  • Complete item includes nylon bushing, U10 Ace-Thred tap, and one #7 nylon adapter
The heavy glass sheath is topped with a 45/50 standard taper joint with a #25 Ace-Thred. Complete unit also includes one #25 nylon bushing with a #7 Ace-Thred tap, and one #7 nylon adapter designed for use with ¼" O.D. probes.
probes within kilo scale reactors.