Cajon Unions, AIRFREE®, Schlenk, Chemglass Inc
Supplier: Chemglass
AF-0067-20 AF-0067-21 AF-0067-22 AF-0067-04 AF-0067-05 AF-0067-06
80060-022EA 6.19 CAD
80060-022 80060-024 80060-026 80060-016 80060-018 80060-020
Cajon Unions, AIRFREE®, Schlenk, Chemglass Inc
Tubing Transfer Tubing
Unions are made from 316 stainless steel.
It provide seals directly from the listed sizes on a glass to glass basis, or it can be used with the cuffs and flexible tubing. They can be used with glass tubing as much as 0.0312" undersize.
Ordering information: Replacement O-Rings made of 70 durometer viton is available and must be ordered separately.