Mudwatt Grow a Living Fuel Cell
Supplier: TRYGVE ENTERPRISES
1081101 1081105 1081106
470225-212EA 81.2 CAD
470225-212 470226-470 470226-472
Mudwatt Grow a Living Fuel Cell
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Create clean energy from mud!
  • Experiment with different soils, and food products to see how the effects power production
  • Great for use in classroom and home school settings
  • Can be used over and over without the need of any batteries
MudWatt engages kids in STEM in a whole new way, using the power of electricity-generating bacteria living in the soil beneath our feet! In a few days, the LED will begin to blink using power generated by your mud. Allowing more time for the bacteria to grow you’ll be able to power a digital clock and thermometer. TheMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable