Bottles, Aluminum, Bürkle
Supplier: BURKLE INC
0327-0600 0300-0327 0327-0300 0327-1200 0327-0038 0327-0060 0327-0120 0327-3000 0327-5600
75997-608EA 12.99 CAD
75997-608 76047-102 76077-114 76077-116 76077-164 76077-166 76077-168 76183-158 76183-160
Bottles, Aluminum, Bürkle
Bottles Metal Bottles
The aluminum bottle is particularly suited for storing, transporting, or sending substances and reference samples which have to be stored completely unaltered
  • Pure aluminum (material AL 99.5)
  • With tamper-evident screw cap made of PP, with aluminum inner seal
  • Suitable for food (exception: highly acidic and salt-containing foods)
  • With UN approval (for 60 to 5600 ml [2.03 to 189.36 oz.])
These bottles can be completely emptied without residues. They are odor-neutral, impervious to light and rustproof.