Sodium phosphate dibasic 1 M in aqueous solution, sterile

Supplier: TEKNOVA CA

Synonyms: di-Sodium hydrogen phosphate anhydrousSodium phosphate dibasicsec-Sodium phosphateDisodium phosphatedi-Sodium hydrogen orthophosphateSodium hydrogen phosphate

S0212 S0212
CA100216-420EA 386.5 CAD
100216-420 CA100216-420
Sodium phosphate dibasic 1 M in aqueous solution, sterile
di-Sodium hydrogen phosphate

Sodium phosphate, dibasic solution may form precipitate, gently warm the solution if precipitated.

Formula: Na₂HPO₄
MW: 141.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR