Manual Syringes for Beckman/Altex, Rheodyne, SSI, and Valco Valves, SGE, Trajan Scientific and Medical
Supplier: SGE (Trajan)
007300 007310 001301 004300 6310 006300 003300 005300
60361-194EA 146.53 CAD
60361-194 60361-150 60361-176 60361-482 60361-470 60361-184 60361-166 60361-468
Manual Syringes for Beckman/Altex, Rheodyne, SSI, and Valco Valves, SGE, Trajan Scientific and Medical
Syringes Syringes for Chromatography
Manual syringes for use with Beckman/Altex, Rheodyne, SSI, and Valco Valves.
  • 5 µL to 500 µL syringe volumes
When using the complete loop fill technique, the syringe capacity should be greater than twice the loop volume
For the partial loop fill technique, the injection volume should be no greater than half the loop capacity. The injection size sets the injection volume.
Note: Users of older Valco injectors requiring 3/4" long needles must fit a Valco VISF-2 adapter.
The loop capacity sets the injection volume.