SP Wilmad-LabGlass Tubing Cutter, SP Industries
Supplier: SP Industries
BP-1928-100
14007-350EA 62.38 CAD
14007-350
SP Wilmad-LabGlass Tubing Cutter, SP Industries
Cutters
This simple guillotine-style polymer tubing cutter provides flat 90°, burr-free cuts for flangeless fittings requiring leak-free seals.
  • Construction is durable
  • Made of molded glass-filled nylon
The cutter is specifically designed to cut PTFE, PEEK, and ETFE tubing in either 1/8 or 1/6" O.D. sizes.