D-(+)-Xylose 98,0 USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

XY110-06 XY110-50KGBL
CA11024-440EA 464 CAD
CA11024-440 75812-620
D-(+)-Xylose 98,0 USP
D(+)-Xylose

XYLOSE, USP is a sugar that occurs in wood and is classified as being an aldopentose type of monosaccharide. It is used in veterinary fields to asses the condition of animals internal organs, notably the good functioning of the kidneys and intestines. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331
Melting Pt: 154
Storage Temperature: Ambient
MDL Number: MFCD00151475
CAS Number: 58-86-6
Merck Index: 13,10142
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Specific Rotation, [A]D25 +18.2 to +19.4
Color of a 10% Solution To Pass Test
Loss On Drying max. 0.1 %
Organic Impurities To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR