Xylitol ≥98.5% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

X1064-12KG X1064-25KGBL
75812-616EA 2737.98 CAD
75812-616 75812-618
Xylitol ≥98.5% FCC
Xylitol

Xylitol, FCC is used as nutritive sweetener as a sugar substitute to produce “sugar-free” foods and is commonly used in the production of chewing gums, mints and other candies. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₅H₁₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS Number: 87-99-0

Order Now

Specification Test Results

Assay (C₅H₁₂O₅) 98.5 - 101.0 %
Lead (Pb) max. 1 mg/kg
Nickel (Ni) max. 1 mg/kg
Other Polyols max. 1.0 %
Reducing Sugars max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR