Urea, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

U1024-12KG U1024-2.5KG U1024-45KGBL U1024-500GM
75812-574EA 2433.04 CAD
75812-574 75812-576 75812-578 75812-580
Urea, BiotechGrade
Urea

Urea, BiotechGrade is an odorless and colorless solid that is an important nitrogen-containing substance found in mammal urine. In concentrations up to 10 M, urea is a powerful protein denaturant as it disrupts the noncovalent bonds in the proteins. This property can be exploited to increase the solubility of some proteins. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197
Melting Pt: 132.7
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless,Transp
Odor Odorless
Assay ([(NH₂)₂Co]) 99.0 - 100.5%
Melting Point 132° - 134°C
Solubility in Water 1 G in 1 ML


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR