Undec-10-enoic acid ≥97.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

UN105-160KGBL UN105-20LT
75812-588EA 33430.35 CAD
75812-588 75812-590
Undec-10-enoic acid ≥97.0% USP
Undec-10-enoic acid

UNDECYLENIC ACID, USP is an organic unsaturated fatty acid that is a derived from castor oil. It is used in pharmaceutical and cosmetics for its antimicrobial properties. It the medical world it is used in over-the-counter products that treat skin disorders such as skin irritations, burning, infections and itching. It is also used in biosensors that are silicon-based. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₁H₂₀O₂
Density: 0.910
Storage Temperature: Ambient
MDL Number: MFCD00004442
CAS Number: 112-38-9

Order Now

Specification Test Results

Assay 97.0 - 100.5%
Specific Gravity 0.910 - 0.913
Congealing Range 21°C min.
Refractive Index 1.447 - 1.448
Iodine Value 131 - 138


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR