Titanium(IV) oxide ≥99.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

T1083-12KGPL T1083-45KGBL
75812-528EA 1660.9 CAD
75812-528 75812-530
Titanium(IV) oxide ≥99.0% FCC
Titanium (IV) oxide

Titanium Dioxide, FCC is used as a food coloring and has an E-number, E171. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Health hazard

Formula: TiO₂
MW: 79.88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5 %
Acid-Soluble Substances max. 0.5 %
Aluminum Oxide and /Or Silicon Dioxide max. 2.0 %
Antimony (Sb) max. 2 mg/kg
Arsenic (As) max. 1 mg/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR