L(+)-Tartaric acid ≥99.7%, granulated FCC

Supplier: Spectrum Chemical Mfg. Corp.

T1009-12KG T1009-25KGBL
75812-518EA 3097.59 CAD
75812-518 75812-520
L(+)-Tartaric acid ≥99.7%, granulated FCC
L(+)-Tartaric acid

Tartaric Acid, Granular, FCC is used as an leavening additive and flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications.


Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₄H₆O₆
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₄H₆O₆; Dried Basis) 99.7 - 100.5 %
Optical Rotation, [A] 25D +12.0° to +13.0
Lead max. 2 mg/kg
Loss On Drying max. 0.5 %
Oxalate To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR