D-(+)-Sucrose, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranosideSucroseD(+)-Saccharoseβ-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranosideα-ᴅ-Glc-(1→2)-β-ᴅ-Fru

S1699-45KGBL S1699-2.5KG S1699-12KG S1699-1KG S1699-500GM
75812-368EA 318.72 CAD
75812-368 75812-366 75812-360 75812-364 75812-362
D-(+)-Sucrose, BiotechGrade
D(+)-Sucrose

Sucrose, BiotechGrade - is a disaccharide composed of glucose and fructose. Sucrose is used for a variety of applications such as a supplement in plant, insect, and bacterial culture media. Spectrum offers highly pure reagents suitable for biotech research and analysis.

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
https://ca.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg

Order Now

Specification Test Results

Appearance White, Colorless
Odor Odorless
Specific Rotation @ 25 C (Dried Basis; 105 C) +65.9 min.
Residue On Ignition max. 0.05%
Chloride (Cl) max. 0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR