Spearmint oil (natural) FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

SP306-100ML SP306-2.5LTGL SP306-25LBBL SP306-500MLGL SP306-50KGBL
75812-480EA 293.82 CAD
75812-480 75812-482 75812-484 75812-486 75812-488
Spearmint oil (natural) FCC
Spearmint oil

Spearmint Oil, Natural, FCC is used for cooking, flavoring, and as a digestive support. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Exclamation mark
UN1197

Density: 0.918
Storage Temperature: Ambient
MDL Number: MFCD00148459
CAS Number: 8008-79-5
UN: 1197
ADR: 3,III

Order Now

Specification Test Results

Assay (Ketones ; V/V) 55.0 % min.
Angular Rotation -48 to -59
Reactivity To Pass Test
Refractive Index @ 20°C 1.484 -1.491
Solubility in Alcohol To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR