D-(-)-Sorbitol solution 70% in aqueous solution ≥70% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
D-Sorbitol
, Sorbitol
, D(-)-Glucitol
, (Sorbitol)
, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
Sorbitol Solution, 70 Percent, FCC is used as a sugar substitute and bonding agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
HOH₂C(CH(OH))₄CH₂OH MW: 182.17 g/mol Density: 1.280 Storage Temperature: Ambient |
MDL Number:
MFCD00004708 CAS Number: 50-70-4 |
Specification Test Results
Assay | 64.0% min. |
pH of a 14 % Solution | 5.0 - 7.5 |
Water | 28.5 - 31.5% |
Lead (Pb) | max. 1 mg/kg |
Nickel (Ni) | max. 1 mg/kg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...