D-(-)-Sorbitol ≥91%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol

S1523-12KG S1523-45KGBL
75812-346EA 1560.25 CAD
75812-346 75812-348
D-(-)-Sorbitol ≥91%, powder FCC
D(-)-Sorbitol

Sorbitol, Powder, FCC is used is as a sugar substitute sweetener in many foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 295
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 91.0 - 100.5%
pH of a 10% Solution 3.5 - 7.0
Lead (Pb) max. 1 mg/kg
Nickel (Ni) max. 1 mg/kg
Reducing Sugars max. 0.3%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR