Sodium thiosulfate pentahydrate ≥99.0%, crystals FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

S1502-12KG S1502-50KGBL
75812-330EA 1907.93 CAD
75812-330 75812-332
Sodium thiosulfate pentahydrate ≥99.0%, crystals FCC
Sodium thiosulphate pentahydrate

Sodium Thiosulfate, Pentahydrate, Crystal, FCC is used as an antioxidant and sequestrant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
Merck Index: 12,08844

Order Now

Specification Test Results

Assay (Dried Basis) 99.0-100.5 %
Water 32.0- 37.0 %
Lead (Pb) max. 2 mg/kg
Selenium (Se) max. 0.003 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR