Saccharin sodium dihydrate ≥98.0%, granulated FCC

Supplier: Spectrum Chemical Mfg. Corp.

S1421-100GM S1421-12KG
75812-304EA 375.17 CAD
75812-304 75812-306
Saccharin sodium dihydrate ≥98.0%, granulated FCC
Saccharin sodium dihydrate

Sodium Saccharin, Dihydrate, Granular, FCC is a white crystalline solid whose most common use is as an artificial sweetener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₇H₅NNaO₃S·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00151213
CAS Number: 6155-57-3

Order Now

Specification Test Results

Assay (Anhydrous Basis) 98.0 - 101.0 %
Alkalinity To Pass Test
Benzoate and Salicylate To Pass Test
Lead (Pb) max. 2 mg/kg
Readily Carbonizable Substances To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR