L(+)-Potassium sodium tartrate tetrahydrate ≥99.0%, granular FCC
Supplier: Spectrum Chemical Mfg. Corp.
Sodium Potassium Tartrate, Tetrahydrate, Granular, FCC is used as a sequestrant and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
KOOC(CHOH)₂COONa·4H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150989 CAS Number: 6381-59-5 |
|
Specification Test Results
Assay | 99.0 - 102.0 % |
Alkalinity | To Pass Test |
Water | 21.0 - 26.0 % |
Lead (Pb) | max. 2 mg/kg |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...